ENA444725430 Enamine

1-(3-fluoropyridin-4-yl)ethan-1-amine

  • MDL number MFCD18655697
  • Cas Number : 1270361-88-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203848

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203848
Availability details for
Quantity
  • Availability
  • Add to Cart