ENA444725431 Enamine

N-(2,2-dimethylpropyl)-N-methylcarbamoyl chloride

  • MDL number MFCD23790236
  • Cas Number : 1543028-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203849

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203849
Availability details for
Quantity
  • Availability
  • Add to Cart