ENA444725434 Enamine

N-(4,4,4-trifluorobutyl)acetamide

  • MDL number MFCD23727915
  • Cas Number : 1546526-96-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203852

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203852
Availability details for
Quantity
  • Availability
  • Add to Cart