ENA444725435 Enamine

2-cyclopropanesulfonamidoacetic acid

  • MDL number MFCD16653438
  • Cas Number : 1203136-68-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203853

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203853
Availability details for
Quantity
  • Availability
  • Add to Cart