ENA444725438 Enamine

2-bromo-6-fluorobenzene-1-thiol

  • MDL number MFCD18089292
  • Cas Number : 59594-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203866

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203866
Availability details for
Quantity
  • Availability
  • Add to Cart