ENA444725439 Enamine

1-tert-butyl-1H-1,2,4-triazol-5-amine

  • MDL number MFCD16845175
  • Cas Number : 1247143-86-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203869

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203869
Availability details for
Quantity
  • Availability
  • Add to Cart