ENA444725451 Enamine

N-[2-(5-chloro-1H-indol-1-yl)ethyl]oxan-4-amine

  • MDL number MFCD19536805
  • Cas Number : 1410957-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203888

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203888
Availability details for
Quantity
  • Availability
  • Add to Cart