ENA444725458 Enamine

2-oxo-3-(1-phenyl-1H-pyrazol-4-yl)propanoic acid

  • MDL number MFCD09734432
  • Cas Number : 1052567-31-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203941

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203941
Availability details for
Quantity
  • Availability
  • Add to Cart