ENA444725461 Enamine

hexahydroimidazo[1,5-c][1,3]thiazole-5,7-dione

  • MDL number MFCD00729065
  • Cas Number : 5718-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203947

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203947
Availability details for
Quantity
  • Availability
  • Add to Cart