ENA444725462 Enamine

5-ethoxy-2-nitrobenzoic acid

  • MDL number MFCD09736050
  • Cas Number : 78361-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203982

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203982
Availability details for
Quantity
  • Availability
  • Add to Cart