ENA444725483 Enamine

spiro[1,3-dioxaindane-2,1'-cyclopentan]-6-ol

  • MDL number MFCD24718516
  • Cas Number : 77377-09-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204008

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204008
Availability details for
Quantity
  • Availability
  • Add to Cart