ENA444725485 Enamine

N-benzyl-2-methoxy-N-methylacetamide

  • MDL number MFCD03576138
  • Cas Number : 139765-14-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204010

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204010
Availability details for
Quantity
  • Availability
  • Add to Cart