ENA444725495 Enamine

ethyl 4-(ethylamino)cyclohexane-1-carboxylate

  • MDL number MFCD12808765
  • Cas Number : 943551-58-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203099

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203099
Availability details for
Quantity
  • Availability
  • Add to Cart