ENA444725497 Enamine

N-cyclohexyl-2-hydroxy-N-phenylacetamide

  • MDL number MFCD08443407
  • Cas Number : 926190-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202876

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202876
Availability details for
Quantity
  • Availability
  • Add to Cart