ENA444725508 Enamine

6-ethyl-3-(propan-2-yl)-[1,2]oxazolo[5,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD12410199
  • Cas Number : 1096805-70-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203115

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203115
Availability details for
Quantity
  • Availability
  • Add to Cart