ENA444725510 Enamine

1-(bromomethyl)-3-propoxybenzene

  • MDL number MFCD11103172
  • Cas Number : 866596-42-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203117

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203117
Availability details for
Quantity
  • Availability
  • Add to Cart