ENA444725517 Enamine

methyl 3-hydroxy-1-methyl-1H-pyrrole-2-carboxylate

  • MDL number MFCD12924083
  • Cas Number : 113602-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203122

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203122
Availability details for
Quantity
  • Availability
  • Add to Cart