ENA444725520 Enamine

4-methyl-2-(1-methyl-1H-pyrazol-4-yl)benzaldehyde

  • MDL number MFCD24238613
  • Cas Number : 1558171-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204130

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204130
Availability details for
Quantity
  • Availability
  • Add to Cart