ENA444725527 Enamine

5-cyclopropyl-5-propylimidazolidine-2,4-dione

  • MDL number MFCD20729148
  • Cas Number : 90565-78-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204144

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204144
Availability details for
Quantity
  • Availability
  • Add to Cart