ENA444725528 Enamine

2-chloro-2-(4-chlorophenyl)acetonitrile

  • MDL number MFCD16146977
  • Cas Number : 89978-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204146

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204146
Availability details for
Quantity
  • Availability
  • Add to Cart