ENA444725529 Enamine

4-chloro-3-hydroxy-1,2-thiazole-5-carboxylic acid

  • MDL number MFCD09743741
  • Cas Number : 948292-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204147

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204147
Availability details for
Quantity
  • Availability
  • Add to Cart