ENA444725537 Enamine

2-oxo-2-(2-oxo-1,2,3,4-tetrahydroquinolin-6-yl)acetic acid

  • MDL number MFCD11103001
  • Cas Number : 1152495-52-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204182

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204182
Availability details for
Quantity
  • Availability
  • Add to Cart