ENA444725541 Enamine

1-(2,5-dimethylphenyl)-2-hydroxyethan-1-one

  • MDL number MFCD08442414
  • Cas Number : 503808-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204187

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204187
Availability details for
Quantity
  • Availability
  • Add to Cart