ENA444725552 Enamine

6H,7H-thieno[3,2-d]pyrimidine-2,4-diol

  • MDL number MFCD13181524
  • Cas Number : 913581-92-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202883

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202883
Availability details for
Quantity
  • Availability
  • Add to Cart