ENA444725553 Enamine

[4-(1,3-dioxaindan-5-yl)butan-2-yl]hydrazine hydrochloride

  • MDL number MFCD01722404
  • Cas Number : 7296-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204252

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204252
Availability details for
Quantity
  • Availability
  • Add to Cart