ENA444725557 Enamine

4-ethynyl-3-fluoropyridine

  • MDL number MFCD13189776
  • Cas Number : 1196156-81-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204291

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204291
Availability details for
Quantity
  • Availability
  • Add to Cart