ENA444725561 Enamine

methyl 2-(3-nitro-1H-1,2,4-triazol-1-yl)acetate

  • MDL number MFCD01027023
  • Cas Number : 70965-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106-108
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204308

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204308
Availability details for
Quantity
  • Availability
  • Add to Cart