ENA444725562 Enamine

6-[(1-methylpiperidin-4-yl)amino]-1,2,3,4-tetrahydroquinolin-2-one

  • MDL number MFCD12507212
  • Cas Number : 1157387-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204310

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204310
Availability details for
Quantity
  • Availability
  • Add to Cart