ENA444725564 Enamine

2-methyl-1-phenyl-1H-1,3-benzodiazole

  • MDL number MFCD00227220
  • Cas Number : 1484-39-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68-72
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204314

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204314
Availability details for
Quantity
  • Availability
  • Add to Cart