ENA444725566 Enamine

2,2-dimethyl-3-phenoxypropan-1-ol

  • MDL number MFCD16302064
  • Cas Number : 1991-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204316

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204316
Availability details for
Quantity
  • Availability
  • Add to Cart