ENA444725571 Enamine

2-(2,3-dioxo-1,2,3,4-tetrahydroquinoxalin-6-yl)-2-oxoacetic acid

  • MDL number MFCD11179620
  • Cas Number : 1094294-14-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204323

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204323
Availability details for
Quantity
  • Availability
  • Add to Cart