ENA444725576 Enamine

(2R)-1-amino-3-methoxypropan-2-ol

  • MDL number MFCD09260573
  • Cas Number : 1174289-21-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204338

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204338
Availability details for
Quantity
  • Availability
  • Add to Cart