ENA444725579 Enamine

3-(diethylsulfamoyl)benzene-1-sulfonyl chloride

  • MDL number MFCD09734390
  • Cas Number : 953884-53-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203230

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203230
Availability details for
Quantity
  • Availability
  • Add to Cart