ENA444725580 Enamine

2-chloro-4-methyl-1,3-thiazole-5-sulfonamide

  • MDL number MFCD06658400
  • Cas Number : 348086-67-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204349

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204349
Availability details for
Quantity
  • Availability
  • Add to Cart