ENA444725583 Enamine

N-ethyl-2-(piperidin-2-yl)acetamide

  • MDL number MFCD11204459
  • Cas Number : 102308-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204352

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204352
Availability details for
Quantity
  • Availability
  • Add to Cart