ENA444725585 Enamine

2-amino-3-methoxybenzene-1-sulfonamide

  • MDL number MFCD18389711
  • Cas Number : 393089-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204357

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204357
Availability details for
Quantity
  • Availability
  • Add to Cart