ENA444725587 Enamine

2-amino-5-(trifluoromethyl)benzene-1-thiol

  • MDL number MFCD15528706
  • Cas Number : 86691-07-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204363

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204363
Availability details for
Quantity
  • Availability
  • Add to Cart