ENA444725588 Enamine

(1S)-1-(3-bromo-4-chlorophenyl)ethan-1-ol

  • MDL number MFCD24232008
  • Cas Number : 1567895-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204365

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204365
Availability details for
Quantity
  • Availability
  • Add to Cart