ENA444725591 Enamine

(2-methylpentan-2-yl)[(thiophen-2-yl)methyl]amine

  • MDL number MFCD28348312
  • Cas Number : 1595634-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204372

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204372
Availability details for
Quantity
  • Availability
  • Add to Cart