ENA444725598 Enamine

[1-(2-methylphenyl)-1H-1,2,3-triazol-4-yl]methanol

  • MDL number MFCD11844961
  • Cas Number : 1096130-75-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204437

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204437
Availability details for
Quantity
  • Availability
  • Add to Cart