ENA444725600 Enamine

6-fluoroquinolin-3-ol

  • MDL number MFCD12405089
  • Cas Number : 1167056-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202891

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202891
Availability details for
Quantity
  • Availability
  • Add to Cart