ENA444725601 Enamine

6-amino-4,5-dichloro-2-methyl-2,3-dihydropyridazin-3-one

  • MDL number MFCD20690927
  • Cas Number : 25717-64-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191-195
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204447

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204447
Availability details for
Quantity
  • Availability
  • Add to Cart