ENA444725606 Enamine

1-(pyridin-2-yl)-1H-1,2,3,4-tetrazol-5-amine

  • MDL number MFCD20700599
  • Cas Number : 76105-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203241

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203241
Availability details for
Quantity
  • Availability
  • Add to Cart