ENA444725607 Enamine

tert-butyl 4-(3-amino-1,1,1-trifluoropropan-2-yl)piperazine-1-carboxylate

  • MDL number MFCD28348203
  • Cas Number : 1698595-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203242

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203242
Availability details for
Quantity
  • Availability
  • Add to Cart