ENA444725608 Enamine

2-methyl-5-[(pyridin-2-yl)methyl]aniline

  • MDL number MFCD21265206
  • Cas Number : 1462272-76-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203243

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203243
Availability details for
Quantity
  • Availability
  • Add to Cart