ENA444725612 Enamine

2'-bromo-[1,1'-biphenyl]-4-carbaldehyde

  • MDL number MFCD05980885
  • Cas Number : 198205-81-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203262

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203262
Availability details for
Quantity
  • Availability
  • Add to Cart