ENA444725615 Enamine

3-{3H-imidazo[4,5-b]pyridin-2-yl}propan-1-amine dihydrochloride

  • MDL number MFCD09029288
  • Cas Number : 1215739-41-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204570

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204570
Availability details for
Quantity
  • Availability
  • Add to Cart