ENA444725653 Enamine

1-(azetidin-3-yl)-3-methylurea hydrochloride

  • MDL number MFCD24593866
  • Cas Number : 1803600-98-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203288

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203288
Availability details for
Quantity
  • Availability
  • Add to Cart