ENA444725655 Enamine

2-hydroxy-N-(pentan-2-yl)propanamide

  • MDL number MFCD12137898
  • Cas Number : 1153455-58-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203292

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203292
Availability details for
Quantity
  • Availability
  • Add to Cart