ENA444725662 Enamine

N-octylacetamide

  • MDL number MFCD00966015
  • Cas Number : 7462-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203300

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203300
Availability details for
Quantity
  • Availability
  • Add to Cart